Product Details

SNP ID
rs35782968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50476567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATGGCGGCAGCCAGCGCTGGGG[C/T]AACCCGGCTGCTCCTGCTCTTGCTG
Phenotype
MIM: 614545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175063.5 180 Missense Mutation GCA,GTA A8V NP_778233.4
NM_206538.3 180 Missense Mutation GCA,GTA A8V NP_996261.1
XM_006723163.3 180 Missense Mutation GCA,GTA A8V XP_006723226.1
XM_011526818.2 180 Missense Mutation GCA,GTA A8V XP_011525120.1
Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 180 UTR 5 NP_001295358.1
NM_138411.2 180 UTR 5 NP_612420.1
XM_011526405.2 180 Intron XP_011524707.2
XM_011526406.2 180 Intron XP_011524708.2

View Full Product Details