Product Details

SNP ID
rs61103451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121651598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCAGAGCCGCACTGGTCTGAG[C/T]GATCGGGTGACACAGGCTTGAATTT
Phenotype
MIM: 610277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORN3 PubMed Links

Gene Details

Gene
MORN3
Gene Name
MORN repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173855.4 887 UTR 3 NP_776254.3
XM_005253869.4 887 UTR 3 XP_005253926.1
XM_011538213.2 887 UTR 3 XP_011536515.1
XM_011538214.2 887 UTR 3 XP_011536516.1
Gene
ORAI1
Gene Name
ORAI calcium release-activated calcium modulator 1
There are no transcripts associated with this gene.

View Full Product Details