Product Details
- SNP ID
-
rs9468061
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:27325699 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCATCTTCCTACCCAATTATCTG[C/T]ACTTTGATATTGATTAATTATGATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VN1R10P
PubMed Links
Gene Details
- Gene
- VN1R10P
- Gene Name
- vomeronasal 1 receptor 10 pseudogene
There are no transcripts associated with this gene.
View Full Product Details