Product Details

SNP ID
rs989702057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53711869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGTGTGATAGAAAATGGGCAT[A/G]AAACCTAAGTGTATGCATGTGTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALCOCO1 PubMed Links

Gene Details

Gene
CALCOCO1
Gene Name
calcium binding and coiled-coil domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143682.1 2295 UTR 3 NP_001137154.1
NM_020898.2 2295 UTR 3 NP_065949.1

View Full Product Details