Product Details

SNP ID
rs58378427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42313793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATAAAGAAAAACAAATTGGAAT[A/G]TAACCAACCAATAATCAAAAGCAAC
Phenotype
MIM: 610750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7851 PubMed Links

Gene Details

Gene
MIR7851
Gene Name
microRNA 7851
There are no transcripts associated with this gene.

Gene
ZCRB1
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033114.3 Intron NP_149105.3
XM_017020124.1 Intron XP_016875613.1

View Full Product Details