Product Details

SNP ID
rs4505290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90955063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGCTCCAGCCCCACGGGTTCCT[A/G]CAGGGGCGGGACCACCTGGCCCAAC
Phenotype
MIM: 611219
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCCD1 PubMed Links

Gene Details

Gene
RCCD1
Gene Name
RCC1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017919.1 Intron NP_001017919.1
NM_033544.2 Intron NP_291022.2
Gene
UNC45A
Gene Name
unc-45 myosin chaperone A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039675.1 Intron NP_001034764.1
NM_001323619.1 Intron NP_001310548.1
NM_001323620.1 Intron NP_001310549.1
NM_001323621.1 Intron NP_001310550.1
NM_018671.4 Intron NP_061141.2
XM_005254963.3 Intron XP_005255020.1
XM_011521779.2 Intron XP_011520081.1
XM_011521780.2 Intron XP_011520082.1
XM_017022407.1 Intron XP_016877896.1

View Full Product Details