Product Details

SNP ID
rs372350604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12170869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCACCACTACACTAAGTCATAC[A/G]CGCTCGGCCACCAGGAGTTGCAGAA
Phenotype
MIM: 609230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT5 PubMed Links

Gene Details

Gene
NUDT5
Gene Name
nudix hydrolase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321647.1 Intron NP_001308576.1
NM_001321648.1 Intron NP_001308577.1
NM_014142.3 Intron NP_054861.2
Gene
SEC61A2
Gene Name
Sec61 translocon alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142627.2 Intron NP_001136099.1
NM_001142628.1 Intron NP_001136100.1
NM_018144.3 Intron NP_060614.2

View Full Product Details