Product Details

SNP ID
rs7684872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99906465 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTCTTTTCAGACAACTCTAATT[A/G]CTTTTCCTGAGCTGAAATTTGCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 Intron NP_001026893.1
NM_001278310.1 Intron NP_001265239.1
NM_001278311.1 Intron NP_001265240.1
XM_011532262.2 Intron XP_011530564.1
XM_011532263.2 Intron XP_011530565.1
XM_011532265.2 Intron XP_011530567.1
XM_017008628.1 Intron XP_016864117.1

View Full Product Details