Product Details

SNP ID
rs56298757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22547516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGTCCCACAGATATCCAGAA[C/G]CCTGACCCTGCCGTGTACCAGCTGA
Phenotype
MIM: 186880
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105370399 PubMed Links

Gene Details

Gene
LOC105370399
Gene Name
uncharacterized LOC105370399
There are no transcripts associated with this gene.

Gene
TRAC
Gene Name
T-cell receptor alpha constant
There are no transcripts associated with this gene.

View Full Product Details