Product Details
- SNP ID
-
hCV25965072
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:64204259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTATTTAATAAATCCATTTCAC[C/T]GAAGTAATGTGTTTCATATTAATAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBE2U
PubMed Links
Gene Details
- Gene
- UBE2U
- Gene Name
- ubiquitin conjugating enzyme E2 U (putative)
There are no transcripts associated with this gene.
View Full Product Details