Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018142.2 | 111 | Intron | NP_060612.2 | ||
XM_005273555.3 | 111 | UTR 5 | XP_005273612.1 | ||
XM_005273556.3 | 111 | Intron | XP_005273613.1 | ||
XM_005273557.3 | 111 | Intron | XP_005273614.1 | ||
XM_006716355.2 | 111 | Intron | XP_006716418.1 | ||
XM_017013606.1 | 111 | Intron | XP_016869095.1 | ||
XM_017013607.1 | 111 | UTR 5 | XP_016869096.1 | ||
XM_017013608.1 | 111 | Intron | XP_016869097.1 | ||
XM_017013609.1 | 111 | Intron | XP_016869098.1 |