Product Details
- SNP ID
-
rs1142889
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:89186405 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTTCCTCCTCCAACTTCTTCTC[G/C]ATTTGGGCCATGTATTCCTGGAAGC
- Phenotype
-
MIM: 612466
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GBP4
PubMed Links
Gene Details
- Gene
- GBP4
- Gene Name
- guanylate binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052941.4 |
1751 |
Missense Mutation |
ATC,ATG |
I545M |
NP_443173.2 |
View Full Product Details