Product Details

SNP ID
rs17758315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133514768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACAGTGGTAGAAGCTGTGGAC[A/G]TAAGTGTAGTCCCAGTCCTGCGGGG
Phenotype
MIM: 615345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM8C PubMed Links

Gene Details

Gene
TMEM8C
Gene Name
transmembrane protein 8C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080483.2 636 Silent Mutation TAC,TAT Y178Y NP_001073952.1

View Full Product Details