Product Details

SNP ID
rs6924483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43507736 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGTTAAGGCAGGTCCTCAGACC[C/T]CAACCTGCCATGCCTTAGGTATGTC
Phenotype
MIM: 610060 MIM: 612658 MIM: 609775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012974.2 Intron NP_001012992.1
NM_001271882.1 Intron NP_001258811.1
Gene
POLR1C
Gene Name
RNA polymerase I subunit C
There are no transcripts associated with this gene.

Gene
TJAP1
Gene Name
tight junction associated protein 1
There are no transcripts associated with this gene.

Gene
YIPF3
Gene Name
Yip1 domain family member 3
There are no transcripts associated with this gene.

View Full Product Details