Product Details
- SNP ID
-
rs76129882
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59639387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGAGCAGTGACAGGCCAAACTA[C/T]GAGAAAAGACAGAGGGAATCAAACT
- Phenotype
-
MIM: 614660
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PATL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77539333] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PATL1
- Gene Name
- PAT1 homolog 1, processing body mRNA decay factor
View Full Product Details