Product Details

SNP ID
rs34135815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1685487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGAGCAATCTTTTTGGAAGTCC[A/G]GAAGAAGCTACTCCTTCCAGCAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRAMP1 PubMed Links

Gene Details

Gene
CRAMP1
Gene Name
cramped chromatin regulator homolog 1
There are no transcripts associated with this gene.

Gene
HN1L
Gene Name
hematological and neurological expressed 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144570.2 129 Silent Mutation CCA,CCG P31P NP_653171.1

View Full Product Details