Product Details
- SNP ID
-
rs200253669
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:17229396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCCAGTCCGCGCCACCGCCGCCG[C/T]CCAGGCCATCGCCACCCTCCGCAGC
- Phenotype
-
MIM: 193060
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VIM
PubMed Links
Gene Details
- Gene
- VIM
- Gene Name
- vimentin
- Gene
- VIM-AS1
- Gene Name
- VIM antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details