Product Details

SNP ID
rs3751440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44022620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCATGAAGACTCTGCTCCTGG[A/G]TCAGGAATGCTTTTTTGTCCCTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00284 PubMed Links

Gene Details

Gene
LINC00284
Gene Name
long intergenic non-protein coding RNA 284
There are no transcripts associated with this gene.

View Full Product Details