Product Details

SNP ID
rs9606756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30610873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTACCATTTTCTTTTCTAAGAA[A/G]TACCAGAGATGGACAGCCATCTGGT
Phenotype
MIM: 605819 MIM: 613441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PES1 PubMed Links

Gene Details

Gene
PES1
Gene Name
pescadillo ribosomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
TCN2
Gene Name
transcobalamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000355.3 316 Missense Mutation ATA,GTA I23V NP_000346.2
NM_001184726.1 316 Missense Mutation ATA,GTA I23V NP_001171655.1

View Full Product Details