Product Details

SNP ID
rs61744161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45785819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCGCTGGGCTTCTCCCCACC[A/G]CTGCCACTGCCGCCACTGCCACCCC
Phenotype
MIM: 605377 MIM: 609857 MIM: 607548
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMPK PubMed Links

Gene Details

Gene
DMPK
Gene Name
dystrophia myotonica protein kinase
There are no transcripts associated with this gene.

Gene
DMWD
Gene Name
dystrophia myotonica, WD repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004943.1 1723 Silent Mutation AGC,AGT S559S NP_004934.1
Gene
RSPH6A
Gene Name
radial spoke head 6 homolog A
There are no transcripts associated with this gene.

View Full Product Details