Product Details

SNP ID
rs145536390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21211964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACTAGATTAAAATCAAATGTAC[C/T]GAAGATATGAGTTAGCAAATACACT
Phenotype
MIM: 164020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPC PubMed Links

Gene Details

Gene
HNRNPC
Gene Name
heterogeneous nuclear ribonucleoprotein C (C1/C2)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077442.1 Intron NP_001070910.1
NM_001077443.1 Intron NP_001070911.1
NM_004500.3 Intron NP_004491.2
NM_031314.2 Intron NP_112604.2
XM_006720125.2 Intron XP_006720188.1
XM_011536708.1 Intron XP_011535010.1
XM_011536709.2 Intron XP_011535011.1
XM_011536710.2 Intron XP_011535012.1
XM_011536711.2 Intron XP_011535013.1
XM_011536712.1 Intron XP_011535014.1
XM_017021252.1 Intron XP_016876741.1
XM_017021253.1 Intron XP_016876742.1
Gene
LINC00641
Gene Name
long intergenic non-protein coding RNA 641
There are no transcripts associated with this gene.

View Full Product Details