Product Details

SNP ID
rs77763564
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.22:30610854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCCCTGTGACCTCATTTGTA[C/T]CATTTTCTTTTCTAAGAAATACCAG
Phenotype
MIM: 605819 MIM: 613441
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PES1 PubMed Links

Gene Details

Gene
PES1
Gene Name
pescadillo ribosomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
TCN2
Gene Name
transcobalamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000355.3 Intron NP_000346.2
NM_001184726.1 Intron NP_001171655.1

View Full Product Details