Product Details

SNP ID
rs574101052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54483841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGTTAAAGTTATGAAACCTAA[C/T]ACCTTTATAATGCATACTGACATTC
Phenotype
MIM: 601713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMFB PubMed Links

Gene Details

Gene
GMFB
Gene Name
glia maturation factor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004124.2 Intron NP_004115.1

View Full Product Details