Product Details

SNP ID
rs74582995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152752356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGGAATTTCCTATACATTTTG[C/T]TGGACAACTAAATAGTATAAATTAT
Phenotype
MIM: 616805 MIM: 192320
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCT1 PubMed Links

Gene Details

Gene
MYCT1
Gene Name
myc target 1
There are no transcripts associated with this gene.

Gene
VIP
Gene Name
vasoactive intestinal peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003381.3 Intron NP_003372.1
NM_194435.2 Intron NP_919416.1
XM_005267135.3 Intron XP_005267192.1
XM_006715562.3 Intron XP_006715625.1

View Full Product Details