Product Details

SNP ID
hCV25968235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145765212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAGGTGCACAGGGCAGCGGGGA[A/G]AGGAGGGCAGGAGTGGAGCCAAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf51 PubMed Links

Gene Details

Gene
C4orf51
Gene Name
chromosome 4 open reading frame 51
There are no transcripts associated with this gene.

Gene
ZNF827
Gene Name
zinc finger protein 827
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306215.1 Intron NP_001293144.1
NM_178835.4 Intron NP_849157.2

View Full Product Details