Product Details

SNP ID
rs73942906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45486384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGTGTCCTGGGCACGATGGTT[A/G]ATCTTTGAGTGAGTTTGCATACGTG
Phenotype
MIM: 126380 MIM: 603183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC1 PubMed Links

Gene Details

Gene
ERCC1
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
There are no transcripts associated with this gene.

Gene
RTN2
Gene Name
reticulon 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005619.4 Intron NP_005610.1
NM_206900.2 Intron NP_996783.1
NM_206901.2 Intron NP_996784.1

View Full Product Details