Product Details

SNP ID
rs7514518
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:157123043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCAAGCACTGTAGTTTAAAAA[A/G]GAAAAAACACCCCCCACCATTAAAA
Phenotype
MIM: 164873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYCSP52 PubMed Links

Gene Details

Gene
CYCSP52
Gene Name
cytochrome c, somatic pseudogene 52
There are no transcripts associated with this gene.

Gene
ETV3
Gene Name
ETS variant 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145312.1 3607 Intron NP_001138784.1
NM_005240.1 3607 Intron NP_005231.1
XM_006711210.2 3607 UTR 3 XP_006711273.1

View Full Product Details