Product Details

SNP ID
rs35530857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10765770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTCCCCCTTTTACGTTCCAC[C/G]TGCGTCTCAGAAATGTCAGACCCTG
Phenotype
MIM: 606505
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC102723313 PubMed Links

Gene Details

Gene
LOC102723313
Gene Name
uncharacterized LOC102723313
There are no transcripts associated with this gene.

Gene
PINX1
Gene Name
PIN2/TERF1 interacting, telomerase inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details