Product Details

SNP ID
hCV25970135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107505239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAATGAAAAAAAACAAAACACA[C/T]GATCAACCTGGAAGAGACAGGAAAT
Phenotype
MIM: 613306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH8 PubMed Links

Gene Details

Gene
ALKBH8
Gene Name
alkB homolog 8, tRNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301010.1 Intron NP_001287939.1
NM_138775.2 Intron NP_620130.2

View Full Product Details