Product Details

SNP ID
rs144436978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71119535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAAGAATCTAAGAAGGAGCAAA[C/G]GCCCTGGGTTGGGAAGACTTATAGG
Phenotype
MIM: 308380 MIM: 300188
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL2RG PubMed Links

Gene Details

Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
There are no transcripts associated with this gene.

Gene
MED12
Gene Name
mediator complex subunit 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005120.2 Intron NP_005111.2

View Full Product Details