Product Details

SNP ID
hCV25971286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126272844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTGACTTACACATGGGTCTTA[C/G]ATTATAGACTTGCAAATAGGACTCC
Phenotype
MIM: 616587 MIM: 613622 MIM: 182180 MIM: 606252
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017547.3 Intron NP_060017.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details