Product Details
- SNP ID
-
rs45618544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:6925014 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATCTTCAGGAAGTTGCATGCTT[G/T]CTGACATGTAATGCTGGCTGCATTA
- Phenotype
-
MIM: 600471
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFA6
PubMed Links
Gene Details
- Gene
- DEFA6
- Gene Name
- defensin alpha 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001926.3 |
|
Intron |
|
|
NP_001917.1 |
View Full Product Details