Product Details

SNP ID
rs113631653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1310356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCCACCTTAAGCAGCAGGCC[A/G]GCTGCAACCCATCATCCGAGGGTGG
Phenotype
MIM: 604942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP1A-SDCBP2 PubMed Links

Gene Details

Gene
FKBP1A-SDCBP2
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SDCBP2
Gene Name
syndecan binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199784.1 1134 UTR 3 NP_001186713.1
NM_015685.5 1134 UTR 3 NP_056500.2
NM_080489.4 1134 UTR 3 NP_536737.3
Gene
SNPH
Gene Name
syntaphilin
There are no transcripts associated with this gene.

View Full Product Details