Product Details

SNP ID
rs116479475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38496544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCCGGATGGTAAGTCTGTGGG[C/T]CCGTCCTCCCTTCGCTGGCCCAGAT
Phenotype
MIM: 602730 MIM: 604051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACVR2B PubMed Links

Gene Details

Gene
ACVR2B
Gene Name
activin A receptor type 2B
There are no transcripts associated with this gene.

Gene
EXOG
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145464.1 Intron NP_001138936.1
NM_001318954.1 Intron NP_001305883.1
NM_001318955.1 Intron NP_001305884.1
NM_001318956.1 Intron NP_001305885.1
NM_001318957.1 Intron NP_001305886.1
NM_001318958.1 Intron NP_001305887.1
NM_001318959.1 Intron NP_001305888.1
NM_005107.3 Intron NP_005098.2
XM_005265633.3 Intron XP_005265690.1
XM_005265635.3 Intron XP_005265692.1
XM_006713442.3 Intron XP_006713505.1
XM_006713444.3 Intron XP_006713507.1
XM_017007584.1 Intron XP_016863073.1
XM_017007585.1 Intron XP_016863074.1
XM_017007586.1 Intron XP_016863075.1
XM_017007587.1 Intron XP_016863076.1
XM_017007588.1 Intron XP_016863077.1
XM_017007589.1 Intron XP_016863078.1
XM_017007590.1 Intron XP_016863079.1
XM_017007591.1 Intron XP_016863080.1
XM_017007592.1 Intron XP_016863081.1
XM_017007593.1 Intron XP_016863082.1
XM_017007594.1 Intron XP_016863083.1

View Full Product Details