Product Details

SNP ID
rs34098002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62605698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCCATCCCACAGGCAGAGCT[G/T]CCGGTCGTGGCCGCAGGTGAGGAAG
Phenotype
MIM: 606374 MIM: 603947 MIM: 180721
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GAT3 PubMed Links

Gene Details

Gene
B3GAT3
Gene Name
beta-1,3-glucuronyltransferase 3
There are no transcripts associated with this gene.

Gene
EML3
Gene Name
echinoderm microtubule associated protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300793.1 2541 Missense Mutation AAG,CAG K621Q NP_001287722.1
NM_001300794.1 2541 Missense Mutation AAG,CAG K620Q NP_001287723.1
NM_153265.2 2541 Missense Mutation AAG,CAG K620Q NP_694997.2
XM_005273878.3 2541 Missense Mutation AAG,CAG K621Q XP_005273935.1
XM_006718489.3 2541 Missense Mutation AAG,CAG K671Q XP_006718552.1
XM_006718490.3 2541 Missense Mutation AAG,CAG K671Q XP_006718553.1
XM_006718491.3 2541 Missense Mutation AAG,CAG K670Q XP_006718554.1
XM_011544896.2 2541 Missense Mutation AAG,CAG K670Q XP_011543198.1
XM_011544897.2 2541 Missense Mutation AAG,CAG K619Q XP_011543199.1
XM_017017480.1 2541 Missense Mutation AAG,CAG K504Q XP_016872969.1
XM_017017481.1 2541 Missense Mutation AAG,CAG K504Q XP_016872970.1
XM_017017482.1 2541 Missense Mutation AAG,CAG K504Q XP_016872971.1
XM_017017483.1 2541 Missense Mutation AAG,CAG K403Q XP_016872972.1
Gene
MTA2
Gene Name
metastasis associated 1 family member 2
There are no transcripts associated with this gene.

Gene
ROM1
Gene Name
retinal outer segment membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details