Product Details

SNP ID
rs17002623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41709509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGGTGGCAGCAGCGACGGCAGC[C/G]GGATGTAGGTGCTGTGCGCGGGATG
Phenotype
MIM: 608797
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MEI1 PubMed Links

Gene Details

Gene
MEI1
Gene Name
meiotic double-stranded break formation protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152513.3 Intron NP_689726.3
XM_011529935.1 Intron XP_011528237.1
XM_011529936.1 Intron XP_011528238.1
XM_011529937.1 Intron XP_011528239.1
XM_011529938.1 Intron XP_011528240.1
XM_011529939.1 Intron XP_011528241.1
XM_011529940.1 Intron XP_011528242.1
XM_011529941.1 Intron XP_011528243.1
XM_011529942.2 Intron XP_011528244.1
XM_011529943.1 Intron XP_011528245.1
XM_011529944.1 Intron XP_011528246.1
XM_011529945.2 Intron XP_011528247.1
XM_011529946.1 Intron XP_011528248.1
XM_011529947.1 Intron XP_011528249.1
XM_011529948.2 Intron XP_011528250.1
XM_011529949.1 Intron XP_011528251.1
XM_011529950.1 Intron XP_011528252.1
XM_011529952.1 Intron XP_011528254.1
XM_011529954.1 Intron XP_011528256.1
XM_011529955.1 Intron XP_011528257.1
XM_011529956.2 Intron XP_011528258.1
XM_017028633.1 Intron XP_016884122.1
XM_017028635.1 Intron XP_016884124.1

View Full Product Details