Product Details

SNP ID
rs148161849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71536121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTTACATTTTTTTGGTTTAC[A/G]TTTCTAACTTTTTGTTTTCCCCCCT
Phenotype
MIM: 300255 MIM: 313650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OGT PubMed Links

Gene Details

Gene
OGT
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181672.2 Intron NP_858058.1
NM_181673.2 Intron NP_858059.1
XM_017029907.1 Intron XP_016885396.1
XM_017029908.1 Intron XP_016885397.1
Gene
TAF1
Gene Name
TATA-box binding protein associated factor 1
There are no transcripts associated with this gene.

View Full Product Details