Product Details
- SNP ID
-
rs139157701
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:55220374 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCTAGGCAGAGCTCTGCAAGGA[C/G]AGGTTGTGTCTTCGTTCTTTCCGCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PAGE5
PubMed Links
Gene Details
- Gene
- PAGE5
- Gene Name
- PAGE family member 5
View Full Product Details