Product Details

SNP ID
rs61733846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63506460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTTGGAGTTGCCCCACTGTCA[T/C]GTCACCTGGAGAAAAACTGGACCCA
Phenotype
MIM: 606096
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LGALS12 PubMed Links

Gene Details

Gene
LGALS12
Gene Name
galectin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142535.1 409 Missense Mutation ACG,ATG T23M NP_001136007.1
NM_001142536.1 409 Missense Mutation ACG,ATG T23M NP_001136008.1
NM_001142537.1 409 Intron NP_001136009.1
NM_001142538.1 409 Intron NP_001136010.1
NM_033101.3 409 Missense Mutation ACG,ATG T23M NP_149092.2
XM_017018458.1 409 Missense Mutation ACG,ATG T23M XP_016873947.1

View Full Product Details