Product Details

SNP ID
rs201543004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13783328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACATCAAGGCATGGCTCTGGTT[C/G]TATATCAAACAATCAGTTATCAGAG
Phenotype
MIM: 300051
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPM6B PubMed Links

Gene Details

Gene
GPM6B
Gene Name
glycoprotein M6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001994.2 Intron NP_001001994.1
NM_001001995.2 Intron NP_001001995.1
NM_001001996.2 Intron NP_001001996.1
NM_001318729.1 Intron NP_001305658.1
NM_005278.4 Intron NP_005269.1
XM_005274489.4 Intron XP_005274546.1
XM_011545497.2 Intron XP_011543799.1
XM_017029432.1 Intron XP_016884921.1
XM_017029433.1 Intron XP_016884922.1
XM_017029434.1 Intron XP_016884923.1

View Full Product Details