Product Details

SNP ID
rs138514927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103347695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTAATCATTAAGGAGCGGAGG[A/C]TTTTGGAGCTGCTAAAATGCCGGAT
Phenotype
MIM: 605502 MIM: 154365 MIM: 604943
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC2 PubMed Links

Gene Details

Gene
DNAJC2
Gene Name
DnaJ heat shock protein family (Hsp40) member C2
There are no transcripts associated with this gene.

Gene
PSMC2
Gene Name
proteasome 26S subunit, ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204453.1 293 UTR 5 NP_001191382.1
NM_002803.3 293 UTR 5 NP_002794.1
XM_005250505.1 293 UTR 5 XP_005250562.1
Gene
SLC26A5
Gene Name
solute carrier family 26 member 5
There are no transcripts associated with this gene.

View Full Product Details