Product Details

SNP ID
rs74416259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17077484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACTGGGACAGGCCCTTCCTGGGG[A/G]GAGAAGCAGTCTCGAGGTGAGGTGG
Phenotype
MIM: 603601 MIM: 180476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3C2A PubMed Links

Gene Details

Gene
PIK3C2A
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha
There are no transcripts associated with this gene.

Gene
RPS13
Gene Name
ribosomal protein S13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017.2 Intron NP_001008.1
Gene
SNORD14A
Gene Name
small nucleolar RNA, C/D box 14A
There are no transcripts associated with this gene.

Gene
SNORD14B
Gene Name
small nucleolar RNA, C/D box 14B
There are no transcripts associated with this gene.

View Full Product Details