Product Details

SNP ID
hCV25981634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90256693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAGAGGCGGCTGCCCGAGGAA[C/T]TGGGACAGCCCTCTTCTCCTTTTGC
Phenotype
MIM: 602706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMC1 PubMed Links

Gene Details

Gene
PSMC1
Gene Name
proteasome 26S subunit, ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002802.2 Intron NP_002793.2

View Full Product Details