Product Details

SNP ID
rs149964516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16121671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACGGTGAGAAAGCCAGGCATCC[C/T]GGGTAGATGCTCCTTGTCTCCTACT
Phenotype
MIM: 165040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB8A PubMed Links

Gene Details

Gene
RAB8A
Gene Name
RAB8A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005370.4 Intron NP_005361.2
XM_011528020.2 Intron XP_011526322.1

View Full Product Details