Product Details

SNP ID
rs4467185
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47319781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCTCTCTCCGATTCCCCTTAG[A/G]ACTCCTTCAATTATACCACCCCTGA
Phenotype
MIM: 113995
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C5AR1 PubMed Links

Gene Details

Gene
C5AR1
Gene Name
complement component 5a receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001736.3 26 Missense Mutation AAC,GAC N2D NP_001727.1
XM_005259190.4 26 UTR 5 XP_005259247.1

View Full Product Details