Product Details

SNP ID
rs11575596
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34662453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCATGTGTTCAGAGGGATCCT[A/G]ACCAAGAACCTCTCTGCCAGACAGC
Phenotype
MIM: 604833 MIM: 600939
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCL27 PubMed Links

Gene Details

Gene
CCL27
Gene Name
C-C motif chemokine ligand 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006664.3 Intron NP_006655.1
Gene
IL11RA
Gene Name
interleukin 11 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142784.2 Intron NP_001136256.1
Gene
LOC730098
Gene Name
uncharacterized LOC730098
There are no transcripts associated with this gene.

View Full Product Details