Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243466.1 | Intron | NP_001230395.1 | |||
NM_001243474.1 | Intron | NP_001230403.1 | |||
NM_001243476.2 | Intron | NP_001230405.1 | |||
NM_052851.2 | Intron | NP_443083.1 | |||
NM_178006.3 | Intron | NP_821074.1 | |||
NM_178007.2 | Intron | NP_821075.1 | |||
XM_011535299.2 | Intron | XP_011533601.1 | |||
XM_017020834.1 | Intron | XP_016876323.1 | |||
XM_017020835.1 | Intron | XP_016876324.1 |