Product Details

SNP ID
rs41269339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:78867695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACGCTACCGGGCTTACGCCACC[C/G]CCTCTCCTCAACACAAGCCCAAACT
Phenotype
MIM: 615375
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRAK1BP1 PubMed Links

Gene Details

Gene
IRAK1BP1
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010844.3 210 Missense Mutation CCC,CGC P40R NP_001010844.1
XM_006715336.3 210 Missense Mutation CCC,CGC P40R XP_006715399.1
XM_011535447.1 210 Missense Mutation CCC,CGC P40R XP_011533749.1
XM_011535448.2 210 Missense Mutation CCC,CGC P40R XP_011533750.1
XM_017010264.1 210 Intron XP_016865753.1
XM_017010265.1 210 Missense Mutation CCC,CGC P40R XP_016865754.1

View Full Product Details