Product Details

SNP ID
rs4706754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:79203152 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATACTATAAGGGCCTTATAAATGC[C/T]AATATTACCTGCTGGCATTATTATA
Phenotype
MIM: 604502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGN3 PubMed Links

Gene Details

Gene
HMGN3
Gene Name
high mobility group nucleosomal binding domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201362.1 Intron NP_001188291.1
NM_001201363.1 Intron NP_001188292.1
NM_001318884.1 Intron NP_001305813.1
NM_001318885.1 Intron NP_001305814.1
NM_001318886.1 Intron NP_001305815.1
NM_001318887.1 Intron NP_001305816.1
NM_001318888.1 Intron NP_001305817.1
NM_004242.3 Intron NP_004233.1
NM_138730.2 Intron NP_620058.1

View Full Product Details