Product Details

SNP ID
rs2664591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45792043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACGCCGAGCAGCCGCGGGGACC[C/T]AGCGGGGCCGAGAGGGGCGGCTTGG
Phenotype
MIM: 611388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP1 PubMed Links

Gene Details

Gene
DNTTIP1
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052951.2 107 Silent Mutation CCC,CCT P13P NP_443183.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 107 Intron NP_542181.1
XM_011528553.2 107 Intron XP_011526855.1
XM_017027668.1 107 Intron XP_016883157.1

View Full Product Details